Läs mer
Tocris Bioscience™ NS 398
Beskrivning
NS 398 is a selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide |
| CAS | 123653-11-2 |
| Analysprocentintervall | >99% |
| Kvantitet | 50 mg |
| Löslighet | Soluble to 100mM in DMSO |
| Molekylformel | C13H18N2O5S |
| Formel vikt | Observed MW: 314.36 |
For Research Use Only