Läs mer
NSC 295642, TRC
Produktkod. 30012875
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012875 5 mg
30003879 10 mg
30012175 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN9005735MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77111-29-6
Molekylformel C14 H15 N4 S2 . Cl . Cu
Molekylvikt (g/mol) 402.42
Synonym NSC 295642, [1-(2-Pyridinyl)ethylidene]-hydrazinecarbodithioic Acid Phenylmethyl Wste Copper Complex
IUPAC-namn copper;(E)-N-[[benzylsulfanyl(sulfido)-$l^{5}-azanylidene]amino]-1-(2-pyridyl)ethanimine;chloride
LEDER [Cl-].[Cu+2].C\C(=N/N=[N]([S-])SCc1ccccc1)\c2ccccn2
Specifikationer
Kemiskt namn eller material [1-(2-Pyridinyl)ethylidene]-hydrazinecarbodithioic Acid Phenylmethyl Wste Copper Complex
Kvantitet 5 mg
InChI formel InChI=1S/C14H14N4S2.ClH.Cu/c1-12(14-9-5-6-10-15-14)16-17-18(19)20-11-13-7-3-2-4-8-13;;/h2-10H,11H2,1H3;1H;/q-1;;+2/p-1/b16-12+;;
Formel vikt 400.9723