Läs mer
Tocris Bioscience™ NSC 617145
Werner syndrome helicase (WRN) inhibitor
Brand: Tocris Bioscience™ 5340/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NSC 617145 is a werner syndrome helicase (WRN) helicase inhibitor (IC50 = 250 nM); acts synergistically with Mitomycin C (Cat. No. 3258) to induce double-strand breaks and chromosomal abnormalities in vitro.Specifikationer
| NSC 617145 | |
| 1,1'-(2,2-Dimethyl-1,3-propanediyl)bis[3,4-dichloro-1H-pyrrole-2,5-dione | |
| Soluble to 20mM in DMSO | |
| Bioactive Small Molecule |
| 203115-63-3 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use