Läs mer
Tocris Bioscience™ NSC 636819
KDM4A/KDM4B inhibitor
Brand: Tocris Bioscience™ 5287/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NSC 636819 is a KDM4A/KDM4B inhibitor; inhibits histone demethylase activity on H3K9me3 and induces apoptosis in LNCaP prostate cancer cells in vitro. Also negatively regulates androgen-responsive genes.Specifikationer
| 1,5-Bis[(1E)-2-(3,4-dichlorophenyl)ethenyl]-2,4-dinitrobenzene | |
| 1618672-71-1 | |
| 50 mg |