Läs mer
Tocris Bioscience™ NSC 668036
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
Brand: Tocris Bioscience™ 5813/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.Specifikationer
| N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
| 144678-63-7 | |
| Soluble to 100mM in DMSO and to 100mM in ethanol | |
| White |