Läs mer
NSC23925, MedChemExpress
Produktkod. 16480288
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16480288 5 mg
16453666 25 mg
16437828 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16480288 Leverantör Medchem Express Leverantörsnummer HY19626
MedChemExpress NSC23925 is a novel, selective and effective P-glycoprotein (Pgp) inhibitor.
NSC23925 is a novel, selective and effective P-glycoprotein (Pgp) inhibitor.
Specifikationer
Kemiskt namn eller material NSC23925
CAS 858474-14-3
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C22H26Cl2N2O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 14.29 mg/mL (33.91 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 4 mg/mL (9.49 mM; ultrasonic and warming and heat to 80°C)
LEDER OC(C1NCCCC1)C2=CC(C3=CC=C(OC)C=C3)=NC4=CC=CC=C24.[H]Cl.[H]Cl
Molekylvikt (g/mol) 421.36
Formel vikt 421.36
Procent renhet 99.23%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death