missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NT5C |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836234
|
Proteintech
16577-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826234
|
Proteintech
16577-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Pyrimidine 5-prime nucleotidase and type II (UMPH2).Specifications
NT5C | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8TCD5, Q9JM14 | |
100360582, 30833, 50773 | |
NT5C Fusion Protein Ag9862 | |
Primary | |
-20°C | |
NT5C |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
5′, 3′ nucleotidase, cytosolic, cdN, Deoxy 5 nucleotidase 1, DNT, dNT 1, DNT1, NT5C, P5N2, PN I, PN II, UMPH2 | |
Nt5c | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |