missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16577-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Pyrimidine 5-prime nucleotidase and type II (UMPH2).Specifications
NT5C | |
Polyclonal | |
Unconjugated | |
NT5C | |
5′, 3′ nucleotidase, cytosolic, cdN, Deoxy 5 nucleotidase 1, DNT, dNT 1, DNT1, NT5C, P5N2, PN I, PN II, UMPH2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100360582, 30833, 50773 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TCD5, Q9JM14 | |
Nt5c | |
NT5C Fusion Protein Ag9862 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |