missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NT5E (Human) Recombinant Protein
Human NT5E full-length ORF ( AAH15940.1, 28 a.a. - 264 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004907-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: ELTILHTNDVHSRLEQTSEDSSKCVNASRCMGGVARLFTKVQQIRRAEPNVLLLDAGDQYQGTIWFTVYKGAEVAHFMNALRYDAMALGNHEFDNGVEGLIEPLLKEAKFPILSANIKAKGPLASQISGLYLPYKVLPVGDEVVGIVGYTSKETPFLSNPGTNLVFEDEITALQPEVDKLKTLNVNKIIALGHSGSEMDKLIAQKVRGVDVVVGGHSNTFLYTGNCFKRIAWARMSR
Specifications
AAH15940.1 | |
5'-nucleotidase, ecto (CD73) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NT5E | |
GST |
4907 | |
Wheat germ expression system | |
25 μg | |
CD73, E5NT, NT, NT5, NTE, eN, eNT | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |