missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15538-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The MW of this protein is 14 kDa, and this antibody specifically recognizes the 14 kDa protein.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.Specifications
NTF2 | |
Polyclonal | |
Unconjugated | |
NUTF2 | |
NTF 2, NTF2, nuclear transport factor 2, NUTF2, Placental protein 15, PP15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10204, 291981, 68051 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61970, P61971, P61972 | |
Nutf2 | |
NTF2 Fusion Protein Ag7879 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |