missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ NTN4 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 15596022
Change view
Click to view available options
Kvantitet:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
15596022 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15596022 Supplier Invitrogen™ Supplier No. PA543391

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: EDVHREKIQL DLEAEFYFTH LIVMFKSPRP AAMVLDRSQD FGKTWKPYKY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

NTN4 contains 3 laminin EGF-like domains, 1 laminin N-terminal domain and 1 NTR domain. NTN4 may play an important role in neural, kidney and vascular development. It promotes neurite elongation from olfactory bulb explants. NTN4 belongs to a family of proteins related to laminins (see LAMA1, MIM 150320) Koch et al. (2000) [PubMed 11038171].[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTN4
Användningsområden Immunohistochemistry, Western Blot
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 2% sucrose and 0.09% sodium azide
Gen NTN4
Genaccessionsnr. Q9HB63, Q9JI33
Gene Alias Beta-netrin; CDY; CDY1A; FLJ23180; FLJ77185; hepar-derived netrin-like protein; netrin 4; netrin 4; netrin-4; netrin-4; Ntn4; PRO3091; RGD1565947
Gensymboler NTN4
Värd art Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NTN4 (aa 110-159).
Reningsmetod Affinity Chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 57764, 59277
Målarter Human, Mouse
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.