Läs mer
Tocris Bioscience™ NTRC 824
Potent and selective non-peptide NTS2 antagonist
Brand: Tocris Bioscience™ 5438/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NTRC 824 is a potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.Specifikationer
| N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine | |
| 1623002-61-8 | |
| 50 mg |