Läs mer
NU 2058, Tocris Bioscience™
Produktkod. 18086022
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18086022 50 mg
18064552 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18086022 Leverantör Tocris Bioscience Leverantörsnummer 3135/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Cdk1 and cdk2 inhibitor
Cyclin-dependent kinase (cdk) 1 and cdk2 inhibitor (Ki values are 5 and 12 μM respectively). Inhibits growth of human tumor cells in vitro (mean GI50 = 13 μM). Also inhibits DNA topoisomerase II ATPase activity (IC50 = 300 μM).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161058-83-9
Molekylformel C12H17N5O
Molekylvikt (g/mol) 247.302
InChI-nyckel MWGXGTJJAOZBNW-UHFFFAOYSA-N
Synonym 6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
PubChem CID 4564
IUPAC-namn 6-(cyclohexylmethoxy)-7H-purin-2-amine
LEDER C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N
Specifikationer
Kemiskt namn eller material NU 2058
Kvantitet 50 mg
Formel vikt 247.3
Procent renhet >99%