Läs mer
NU2058, MedChemExpress
Produktkod. 16477718
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16477718 10 mg
16437586 100 mg
16423697 50 mg
16410188 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16477718 Leverantör Medchem Express Leverantörsnummer HY19316
MedChemExpress NU2058 (O6-(Cyclohexylmethyl)guanine) is a potent, competitive and guanine-based CDK inhibitor with IC50s of 17 μM and 26 μM for CDK2 and CDK1. NU2058 has anti-cancer activity.
NU2058 (O6-(Cyclohexylmethyl)guanine) is a potent, competitive and guanine-based CDK inhibitor with IC50s of 17 μM and 26 μM for CDK2 and CDK1. NU2058 has anti-cancer activity.
Specifikationer
Kemiskt namn eller material NU2058
Fysisk form Solid
CAS 161058-83-9
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C12H17N5O
För användning med (applikation) Cancer-Kinase/protease
Synonym O6-(Cyclohexylmethyl)guanine
Löslighetsinformation DMSO : ≥ 32 mg/mL (129.40 mM)
LEDER NC1=NC(OCC2CCCCC2)=C3NC=NC3=N1
Molekylvikt (g/mol) 247.3
Formel vikt 247.3
Procent renhet 98.78%