Läs mer
NU6140, MedChemExpress
Produktkod. 16446827
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16446827 5 mg
1 options
Produktkod. 16446827 Leverantör Medchem Express Leverantörsnummer HY107419
MedChemExpress NU6140 is a selective CDK2-cyclin A inhibitor (IC50, 0.41 μM), exhibits 10- to 36-fold selectivity over other CDKs. NU6140 also potently inhibits Aurora A and Aurora B, with IC50s of 67 and 35 nM, respectively. Enhances the apoptotic effect, with anti-cancer activity.
NU6140 is a selective CDK2-cyclin A inhibitor (IC50, 0.41 μM), exhibits 10- to 36-fold selectivity over other CDKs. NU6140 also potently inhibits Aurora A and Aurora B, with IC50s of 67 and 35 nM, respectively. Enhances the apoptotic effect, with anti-cancer activity.
Specifikationer
Kemiskt namn eller material NU6140
Fysisk form Solid
CAS 444723-13-1
Färg Light Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H30N6O2
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : 250 mg/mL (591.69 mM; Need ultrasonic)
LEDER CCN(CC)C(C(C=C1)=CC=C1NC2=NC(OCC3CCCCC3)=C4C(N=CN4)=N2)=O
Molekylvikt (g/mol) 422.52
Formel vikt 422.52
Procent renhet 98.03%