Läs mer
Nuarimol, TRC
Produktkod. 30012612
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012612 5 mg
30009967 25 mg
30003709 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012612 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN9253205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63284-71-9
Molekylformel C17 H12 Cl F N2 O
Molekylvikt (g/mol) 314.74
Synonym α-(2-Chlorophenyl)-α-(4-fluorophenyl)-5-pyrimidinemethanol, (±)-Nuarimol, EL 228, EL 228 (fungicide), EL 2289, Nuarimol, Tridal, Trimidal, Trimidal EC, Trimifruit SC
IUPAC-namn (2-chlorophenyl)-(4-fluorophenyl)-pyrimidin-5-ylmethanol
LEDER OC(c1ccc(F)cc1)(c2cncnc2)c3ccccc3Cl
Specifikationer
Kemiskt namn eller material Nuarimol
Kvantitet 5 mg
InChI formel InChI=1S/C17H12ClFN2O/c18-16-4-2-1-3-15(16)17(22,13-9-20-11-21-10-13)12-5-7-14(19)8-6-12/h1-11,22H
Formel vikt 314.0622