missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi Kits

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Kit represents the new generation of anion exchanger. The optimized design results in high yield and ultra-fast processing.

Brand:  Macherey-Nagel™ 740414.2

Additional Details : Weight : 0.30000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 11985442

  • 362.00 SEK / 2 reactions

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Features

  • Optimized column design—Silica resin packed between filters, with prep in just 75 minutes
  • Column filter included—High filter flow rates, parallel lysate clearing, and loading onto the column
  • Improved silica material—Typical yields of 1,000μg and can be used over a wide pH range
  • Proven anion-exchange technology—Transfection-grade plasmid DNA
  • Kits contain: NucleoBond™ Xtra Maxi columns and filter, RNase A, buffers (RES, LYS, NEU, EQU, WASH, ELU) and plastic washers

Applications

  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications

Specifications

Anion Exchange Chromatography Technology
A260/A280:1.80–1.95
Yes
Yes
<600mL (high copy), <1200mL (low copy)
2
<300kbp
Maxi Gravity Flow Column
75 min./prep
1000μg
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <

SDS
Documents
Certificates