Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-Up
Product Code. 15106456
Change view
Click to view available options
No. of Reactions:
12Prep
Unit Size:
12 preps
Product Code. No. of Reactions Unit Size
15106456 12Prep 12 preps
1 options
Product Code. 15106456 Supplier Macherey-Nagel™ Supplier No. 740609.12C
  • ONLINE EXCLUSIVE PRICE
    637.00 SEK
    204.65 SEK / Paket med 12
    Save 432.35 SEK
    67% Av
 Restricted Item
Beräknad leverans: 09-12-2026
Specifications
No. of Reactions 12Prep
Product Type NucleoSpin Gel and PCR Clean-Up
compliance-icons
Product Identifier
  • NucleoSpin Gel and PCR Clean-up
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate