missing translation for 'onlineSavingsMsg'
Learn More

NUDT16 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12889-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880834

  • 3665.00 SEK / 150µL
Estimated Shipment: 23-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RNA-binding and decapping enzyme that catalyzes the cleavage of the cap structure of snoRNAs and mRNAs in a metal-dependent manner. Part of the U8 snoRNP complex that is required for the accumulation of mature 5.8S and 28S rRNA. Has diphosphatase activity and removes m7G and/or m227G caps from U8 snoRNA and leaves a 5′monophosphate on the RNA. Catalyzes also the cleavage of the cap structure on mRNAs. Does not hydrolyze cap analog structures like 7-methylguanosine nucleoside triphosphate (m7GpppG). Also hydrolysis m7G- and m227G U3-capped RNAs but with less efficiencies. Has broad substrate specificity with manganese or cobalt as cofactor and can act on various RNA species. Binds to the U8 snoRNA; metal is not required for RNA-binding. May play a role in the regulation of snoRNAs and mRNAs degradation. Acts also as a phosphatase; hydrolyzes the non-canonical purine nucleotides inosine diphosphate (IDP) and deoxyinosine diphosphate (dITP) as well as guanosine diphosphate (GDP), deoxyguanosine diphosphate (dGDP), xanthine diphosphate (XDP), inosine triphosphate (ITP) and deoxyinosine triphosphate (ITP) to their respective monophosphate derivatives and does not distinguish between the deoxy- and ribose forms. The order of activity with different substrates is IDP > dIDP >> GDP = dGDP > XDP = ITP = dITP. Binds strongly to GTP, ITP and XTP. Participates in the hydrolysis of dIDP/IDP and probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.
Specifications

Specifications

NUDT16
Polyclonal
Unconjugated
NUDT16
Nudix motif 16, NUDT16, U8 snoRNA binding protein H29K, U8 snoRNA decapping enzyme
Rabbit
Antigen Affinity Chromatography
RUO
131870
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96DE0
NUDT16
NUDT16 Fusion Protein Ag3925
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers