missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10484-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the MutT family of nucleotide pyrophosphatases, a subset of the larger NUDIX hydrolase family. The gene product possesses a modification of the MutT sequence motif found in certain nucleotide pyrophosphatases. The enzyme asymmetrically hydrolyzes Ap4A to yield AMP and ATP and is responsible for maintaining the intracellular level of the dinucleotide Ap4A, the function of which has yet to be established. This gene may be a candidate tumor suppressor gene. Alternative splicing has been observed at this locus and three transcript variants, all encoding the same protein, have been identified.Specifications
NUDT2 | |
Polyclonal | |
Unconjugated | |
NUDT2 | |
Ap4A hydrolase, Ap4Aase, APAH1, Diadenosine tetraphosphatase, Nudix motif 2, NUDT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297998, 318, 66401 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50583, P56380, Q6PEC0 | |
NUDT2 | |
NUDT2 Fusion Protein Ag0743 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |