missing translation for 'onlineSavingsMsg'
Learn More

NUDT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20542-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840095

  • 1236.20 SEK / 20µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 ).
Specifications

Specifications

NUDT3
Polyclonal
Unconjugated
NUDT3
DIPP, DIPP 1, DIPP1, Nudix motif 3, NUDT3
Rabbit
Antigen Affinity Chromatography
RUO
11165, 56409
-20°C
Liquid
Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95989, Q9JI46
Nudt3
NUDT3 Fusion Protein Ag14587
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers