missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMBL Mouse anti-Human, Clone: 5F6E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66155-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.Specifications
NUMBL | |
Monoclonal | |
1.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y6R0 | |
NUMBL | |
NUMBL Fusion Protein Ag16771 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
5F6E1 | |
Unconjugated | |
NUMBL | |
CAG3A, CTG3a, NBL, numb homolog (Drosophila) like, Numb like protein, NUMB R, Numb related protein, NUMBL, NUMBLIKE, NUMBR, TNRC23 | |
Mouse | |
Protein A | |
RUO | |
9253 | |
-20°C | |
Liquid |