Läs mer
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, 1.8mL, 49mm, internal thread
Nunc Cryotubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage applications.
Brand:  Thermo Scientific™ 368632
Produktkod. 11391645
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
Thermo Scientific™ Nunc Cryotubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage applications.
  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications