Learn More
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
Brand: Thermo Scientific™ 362697_OLDNS
Additional Details : Weight : 1.65000kg
Description
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
Features:
- Integrated leakproof cap prevents cross-contamination and lost closures
- Clarified polypropylene combines chemical resistance and high-speed tolerances
- Writing area and flat caps for sample identification
- Volume markings and graduations
- DNase/RNase free
- Product raw materials are biocompatible per USP Class VI test
- Product is non-cytotoxic and non-pyrogenic
- Available in sterilized (SAL 10-6)
- Max RCF: 8500xg for the fixed-angle rotor, 7000xg for swing bucket rotor***
- Operating Temperature: -20 C to 37C
- Shelf life: 2 years
***When fully supported by conical rotor cavity or conical adaptor
Ordering Information:
Tubes are supplied in a bulk pack (sleeve) or a rack.