Learn More
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes
Open and close Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes with one hand due to a proprietary hinged-cap system.
108.94 SEK - 3679.15 SEK
Specifications
Material PP (Polypropylene)
No. per Case 500
View More Specs
Products 7
Product Code Brand Capacity (Metric) Max. Force Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Max. Force Packaging Price Quantity & Availability  
11301645 View Documents Thermo Scientific™
362695
15mL 8,500 x g Racked 262.80 SEK
Pack of 50 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 1
Add to basket
10571663 View Documents Thermo Scientific™
362697
50mL 9,500 x g Racked 3679.15 SEK
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
18-06-2020
Add to basket
12195700 View Documents Thermo Scientific™
362694
15mL 8,500 x g Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10777414 View Documents Thermo Scientific™
362695
15mL 8,500 x g Racked This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11311645 View Documents Thermo Scientific™
362696
50mL 9,500 x g Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10048631 View Documents Thermo Scientific™
362696
50mL 9,500 x g Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10512802 View Documents Thermo Scientific™
362694
15mL 8,500 x g Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Features:
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Conical bottom
  • Clarified polypropylene combines chemical resistance and high-speed tolerances
  • Use 50mL tubes up to 9500 x g; 15mL tubes withstand forces up to 8500 x g
  • White writing area and flat caps for sample identification
  • Printed volume markings and graduations
  • Standard dimensions
Ordering Information:
Tubes are supplied in a bulk pack (sleeve) or a rack.
Warranty: 90 days
Specifications
Specifications
PP (Polypropylene)
500
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.