Läs mer
Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes
Beskrivning
Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies with Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes.
- Prevents attachment of adherent cells to the surface
- Ideal for culturing embryroid bodies, neurospheres or other anchorage-independent cell clusters
- Sterilized by gamma-irradiation and certified to be non-pyrogenic
Includes: Lid
Specifikationer
Specifikationer
| Förslutning ingår | Closure Included |
| Stängningsmaterial | Polystyrene |
| Beskrivning | Multidish |
| Bottenform | Flat |
| Kapacitet (metrisk) | 1 mL (Well) |
| Material | Polystyrene |
| Sterilitet | Sterile |
| Ytbehandling | Nunclon Delta |
| Typ | Multi Dish |
| Volym (metrisk) Fungerar | 1 ml |
| Visa mer |