Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: White; Sterility: Sterile; Lid: With Lid
3780.00 SEK - 9405.00 SEK
Specifications
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
Lid | With Lid |
Array | 8 x 12 |
Product Code | Brand | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | ||||
10072151
![]() |
Thermo Scientific™
136101 |
96 | 1 Microplate | 50 Microplates |
3780.00 SEK
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||
10182831
![]() |
Thermo Scientific™
136102 |
96 | 10 Microplates | 160 Microplates |
9405.00 SEK
Pack of 160 |
In Stock
Log in to see stock. |
||||
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.
- Background, autoflorescence and crosstalk are improved when you use Nunc White microplates.
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250ÎĽL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
8 x 12 | |
White | |
Low Flange Pinchbar | |
400 ÎĽL | |
50 to 200 ÎĽL | |
Nunclon Delta |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
With Lid | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Sterile | |
Flat Bottom (F-well) |