missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP107 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19217-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.Specifications
NUP107 | |
Polyclonal | |
Unconjugated | |
NUP107 | |
107 kDa nucleoporin, nucleoporin 107kDa, Nucleoporin Nup107, NUP107, NUP84 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103468, 116555, 57122 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52590, P57740, Q8BH74 | |
NUP107 | |
NUP107 Fusion Protein Ag5250 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |