missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP160 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16084-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
NUP160 is 1 of up to 60 proteins that make up the 120-MD nuclear pore complex, which mediates nucleoplasmic transport.Specifications
NUP160 | |
Polyclonal | |
Unconjugated | |
NUP160 | |
160 kDa nucleoporin, KIAA0197, nucleoporin 160kDa, Nucleoporin Nup160, NUP120, NUP160 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23279, 311182, 59015 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12769, Q9Z0W3 | |
Nup160 | |
NUP160 Fusion Protein Ag9083 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |