Learn More
NUP50 Mouse anti-Human, Unconjugated, Clone: 1D5A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67001-1-IG-150UL
 View more versions of this product
Product Code. 16879355
  • 4652.90 SEK / 150µL
Description
The NUP50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUP50. This antibody recognizes human antigen. The NUP50 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NUP50
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D5A7
Unconjugated
150 μL
10762
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
NUP50 Mouse anti-Human, Unconjugated, Clone: 1D5A7, Proteintech > 150 μL