missing translation for 'onlineSavingsMsg'
Learn More

NUP98-NUP96 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12329-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16869983

  • 4721.67 SEK / 150µL
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody was raised against the carboxy-terminal region of Nup98-Nup96, it recognizes all Nup98-Nup96 proteins except Nup98-N. Since the autoproteolysis is very rapid this antibody usually detected 98-115 kDa Nup96 protein in multiple cell lines. It

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications

Specifications

NUP98-NUP96
Polyclonal
Unconjugated
NUP98
96 kDa nucleoporin, ADAR2, ADIR2, Nucleoporin Nup96, NUP196, NUP96, NUP98 NUP96, NUP98-NUP96
Rabbit
Antigen Affinity Chromatography
RUO
269966, 4928, 81738
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49793, P52948, Q6PFD9
NUP98
NUP98-NUP96 Fusion Protein Ag3016
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers