Läs mer
Tocris Bioscience™ Nutlin 3a
Beskrivning
Nutlin 3a is an MDM2 antagonist (IC50 = 90 nM); inhibits the MDM2-p53 interaction. Active enantiomer of Nutlin-3 (Cat. No. 3984). Induces expression for P53 regulated genes. Suppresses tumor growth in vivo >98% in prostate cancer and osteosarcoma cancer models. Antiproliferative.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-piperazinone |
| CAS | 675576-98-4 |
| Analysprocentintervall | >97% |
| Kvantitet | 25 mg |
| Molekylformel | C30H30Cl2N4O4 |
| Formel vikt | Observed MW: 581.49 |