missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Nuts for Fittings
Gives low dead volume connections for optimum chromatography.
Brand: Trajan Scientific and Medical™ 103406
Additional Details : Weight : 0.80000kg
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
1.59 mm x 1.2 mm Hole (Extended) | |
Nickel-plated Brass | |
Nuts for Fittings 1/16 in. x 1.2 mm Hole (Extended) | |
5/Pk. |