Learn More
Trajan Scientific and Medical™ Nuts for Fittings
Gives low dead volume connections for optimum chromatography.
Brand: Trajan Scientific and Medical™ 1034050
Additional Details : Weight : 0.01000kg
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
3.17 mm Hole (Extended) | |
Stainless Steel | |
Nuts for Fittings 1/8 in. Hole (Extended) | |
5/Pk. |