Läs mer
NVP-BKM 120, TRC
Produktkod. 30413000
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413000 10 mg
30411624 5 mg
30412764 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30413000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN92576010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 944396-07-0
Molekylformel C18H21F3N6O2
Molekylvikt (g/mol) 410.39
Synonym BKM 120, 5-(2,6-di-4-Morpholinyl-4-pyrimidinyl)-4-(trifluoromethyl)-2-pyridinamine, NVP-BKM 120
IUPAC-namn 5-(2,6-dimorpholin-4-ylpyrimidin-4-yl)-4-(trifluoromethyl)pyridin-2-amine
LEDER Nc1cc(c(cn1)c2cc(nc(n2)N3CCOCC3)N4CCOCC4)C(F)(F)F
Specifikationer
Kemiskt namn eller material Buparlisib
Kvantitet 10 mg
InChI formel InChI=1S/C18H21F3N6O2/c19-18(20,21)13-9-15(22)23-11-12(13)14-10-16(26-1-5-28-6-2-26)25-17(24-14)27-3-7-29-8-4-27/h9-11H,1-8H2,(H2,22,23)
Formel vikt 410.1678