missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16128-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to NXN.
This gene encodes a member of the thioredoxin superfamily, a group of small, multifunctional redox-active proteins. Members of this family are characterized by a conserved active motif called the thioredoxin fold that catalyzes disulfide bond formation and isomerization. The encoded protein acts a redox-dependent regulator of the Wnt signaling pathway and is involved in cell growth and differentiation.Specifications
NXN | |
Polyclonal | |
Unconjugated | |
NXN | |
NRX, nucleoredoxin, NXN, TRG 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18230, 360577, 64359 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97346, Q6DKJ4 | |
Nxn | |
NXN Fusion Protein Ag9113 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |