missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NY-ESO-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19521-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.Specifications
NY-ESO-1 | |
Polyclonal | |
Unconjugated | |
CTAG1A | |
Cancer/testis antigen 1, cancer/testis antigen 1A, Cancer/testis antigen 6.1, CT6.1, CTAG, CTAG1, CTAG1A, CTAG1B, ESO1, L antigen family member 2, LAGE 2, LAGE2, LAGE2A, LAGE2B, NY-ESO-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246100 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78358 | |
CTAG1A | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |