Learn More
Masterflex™ Nylon Female Luer Fitting
Brand:  Masterflex™ 45502-52
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15171415
  • 156.00 SEK / Pack of 25
Estimated Shipment
28-08-2020
Add to basket
Description
Description
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
Specifications
Specifications
Nylon
Female Luer Fitting
Fits Female luer x 3/32 in. hose barb
25 Pack