Learn More
Cole-Parmer™ Nylon Female Luer Fitting
Brand:  Cole-Parmer™ 45502-12
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 15191415
  • 190.00 SEK / Pack of 25
Estimated Shipment
27-03-2020
Add to basket
Specifications
Specifications
Nylon
Female Luer Fitting
Fits Female luer to 1/16 in. low-profile semi-rigid tubing barb
25 Pack