Learn More
Cole-Parmer™ Nylon Manifold, 10 outlets
Expand your flow and line capacity
Brand:  Cole-Parmer™ 31522-59
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11735238
Quantity Price
1 930.00 SEK / Each
10 875.50 SEK / Each
Estimated Shipment
26-04-2021
Description
Description
  • Constructed of nylon for economic general applications.
  • Offers good chemical compatibility.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
180°F
200 psi
10.75 in.
Nylon Manifold, 10 Outlets
-54°C
1/2 in. NPT(F)
3.81 cm
82°C
Nylon
27.31 cm
-60°F
1/2 in.
3/8 in. NPT(F)
3.81 cm