Learn More
Cole-Parmer™ Nylon Manifold, 4 outlets
Expand your flow and line capacity
Brand:  Cole-Parmer™ 31522-39
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11795218
  • 471.00 SEK / Each
Estimated Shipment
15-06-2021
Add to basket
Description
Description
  • Constructed of nylon for economic general applications.
  • Offers good chemical compatibility.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
180°F
200 psi
4 1/8 in.
Nylon Manifold, 4 Outlets
-54°C
3/8 in. NPT(F)
3.18 cm
3.18 cm
82°C
Nylon
10.48 cm
-60°F
3/8 in.
1/4 in. NPT(F)
1.25 in.
1.25 in.