Learn More
Masterflex™ Nylon Threaded Plug
Threaded fittings offer a more permanent installation. Available in a variety of materials and configurations.
281.00 SEK - 377.00 SEK
Specifications
Product Type | Threaded Plug |
---|---|
Description | NPT(M) Connection |
Autoclavable | Not Autoclavable |
Material | Nylon |
Min. Operating Temperature (English) | -65°F |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11705119
|
Masterflex™
MFLX30401-36 |
1 in. |
377.00 SEK
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11790379
|
Masterflex™
30401-46 |
3/4 in. |
296.00 SEK
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11790369
|
Masterflex™
30401-38 |
1/2 in. |
281.00 SEK
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Nylon threaded plug
- Use with rigid tubing for high-pressure applications
- Very good chemical resistance
- Rated to 200 psi at 77°F (25°C)
- Usable from -65 to +190°F (-53 to +87°C)
Specifications
Threaded Plug | |
Not Autoclavable | |
-65°F | |
200 psi | |
87°C |
NPT(M) Connection | |
Nylon | |
-53°C | |
190°F |