Läs mer
Nystose Trihydrate, TRC
Produktkod. 30135320
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135320 100 mg
30135040 250 mg
17928684 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135320 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN489153100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139523-49-2
Molekylformel C24H42O21 . 3(H 2O)
Molekylvikt (g/mol) 720.64
Synonym α-D-Glucopyranoside, O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-β-D-fructofuranosyl, hydrate (1:3), α-D-Glucopyranoside, O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-β-D-fructofuranosyl, trihydrate (9CI)
IUPAC-namn (2R,3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-2-((((2R,3S,4S,5R)-2-((((2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol trihydrate
LEDER OC[C@]1(O[C@@H]([C@H]([C@@H]1O)O)CO)OC[C@]2(O[C@@H]([C@H]([C@@H]2O)O)CO)OC[C@]3(O[C@@H]([C@H]([C@@H]3O)O)CO)O[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO.O.O.O
Specifikationer
Kemiskt namn eller material Nystose Trihydrate
Kvantitet 100 mg
InChI formel InChI=1S/C24H42O21.3H2O/c25-1-8-12(30)16(34)17(35)21(41-8)45-24(20(38)15(33)11(4-28)44-24)7-40-23(19(37)14(32)10(3-27)43-23)6-39-22(5-29)18(36)13(31)9(2-26)42-22;;;/h8-21,25-38H,1-7H2;3*1H2/t8-,9-,10-,11-,12-,13-,14-,15-,16+,17-,18+,19+,20+,21-,22-,23-,24+;;;/m1.../s1
Formel vikt 720.25