Läs mer
O-(2-Acetamido-2-deoxy-D-glucopyranosylidene)amino N-Phenylcarbamate, TRC
Produktkod. 30132725
Change view
Kvantitet:
5 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132725 100 mg
30005616 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA157250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 132489-69-1
Molekylformel C15 H19 N3 O7
Molekylvikt (g/mol) 353.33
IUPAC-namn [(Z)-[(3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-ylidene]amino] N-phenylcarbamate
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O/C/1=N\OC(=O)Nc2ccccc2
Specifikationer
Kemiskt namn eller material O-(2-Acetamido-2-deoxy-D-glucopyranosylidene)amino N-Phenylcarbamate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C15H19N3O7/c1-8(20)16-11-13(22)12(21)10(7-19)24-14(11)18-25-15(23)17-9-5-3-2-4-6-9/h2-6,10-13,19,21-22H,7H2,1H3,(H,16,20)(H,17,23)/b18-14-/t10-,11-,12-,13-/m1/s1
Formel vikt 353.1223
Procent renhet >95%