Läs mer
O-2-Methoxyethylhydroxylamine, TRC
Produktkod. 30481471
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481471 5 mg
30468263 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481471 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5546205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54149-39-2
Molekylformel C3H9NO2
Molekylvikt (g/mol) 91.11
Synonym O-(2-Methoxyethyl)hydroxylamine
LEDER COCCON
Specifikationer
Kemiskt namn eller material O-2-Methoxyethylhydroxylamine
Kvantitet 5 mg
InChI formel InChI=1S/C3H9NO2/c1-5-2-3-6-4/h2-4H2,1H3