Läs mer
O-Acetyl Abacavir, TRC
Produktkod. 30320200
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30320200 10 mg
1 options
Produktkod. 30320200 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA41677510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Acetylabacavir
CAS 178456-36-5
Molekylformel C16 H20 N6 O2
Kvantitet 10 mg
Synonym 2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, 1-acetate, (1S,4R)-, 2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, acetate (ester), (1S,4R)- (9CI), 2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, acetate (ester), (1S-cis)-, O-Acetylabacavir
InChI formel InChI=1S/C16H20N6O2/c1-9(23)24-7-10-2-5-12(6-10)22-8-18-13-14(19-11-3-4-11)20-16(17)21-15(13)22/h2,5,8,10-12H,3-4,6-7H2,1H3,(H3,17,19,20,21)/t10-,12+/m1/s1
LEDER CC(=O)OC[C@H]1C[C@H](C=C1)n2cnc3c(NC4CC4)nc(N)nc23
IUPAC-namn [(1S,4R)-4-[2-amino-6-(cyclopropylamino)purin-9-yl]cyclopent-2-en-1-yl]methyl acetate
Molekylvikt (g/mol) 328.37
Formel vikt 328.1648