missing translation for 'onlineSavingsMsg'
Läs mer

O-Acetyl-L-serine, TRC

Produktkod. 30410816
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410816 500 mg
30411926 250 mg
30414028 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30410816 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO298210500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5147-00-2
Molekylformel C5H9NO4
Molekylvikt (g/mol) 147.13
Synonym (S)-3-Acetoxy-2-aminopropanoic Acid, NSC 226230, O-Acetylserine
IUPAC-namn (2S)-3-acetyloxy-2-aminopropanoic acid
LEDER CC(=O)OC[C@H](N)C(=O)O

Specifications

Kemiskt namn eller material O-Acetyl-L-serine
InChI formel InChI=1S/C5H9NO4/c1-3(7)10-2-4(6)5(8)9/h4H,2,6H2,1H3,(H,8,9)/t4-/m0/s1
Kvantitet 500 mg
Formel vikt 147.0532
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.