Läs mer
O-Acetyl Lacosamide, TRC
Produktkod. 30138754
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138754 100 mg
30006647 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA179900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1318777-54-6
Molekylformel C14 H18 N2 O4
Molekylvikt (g/mol) 278.3037
Synonym (2R)-2-(Acetylamino)-3-(benzylamino)-3-oxopropyl Acetate, (2R)-2-(Acetylamino)-3-(acetyloxy)-N-(phenylmethyl)propanamide, Lacosamide Imp. B (Pharmeuropa)
IUPAC-namn [(2R)-2-acetamido-3-(benzylamino)-3-oxopropyl] acetate
LEDER CC(=O)N[C@H](COC(=O)C)C(=O)NCc1ccccc1
Specifikationer
Kemiskt namn eller material (2R)-2-(Acetylamino)-3-(benzylamino)-3-oxopropyl Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H18N2O4/c1-10(17)16-13(9-20-11(2)18)14(19)15-8-12-6-4-3-5-7-12/h3-7,13H,8-9H2,1-2H3,(H,15,19)(H,16,17)/t13-/m1/s1
Formel vikt 278.1267
Procent renhet >95%