Läs mer
O-Acetyl Mandelic Acid, TRC
Produktkod. 30328459
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328459 100 mg
1 options
Produktkod. 30328459 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA185963100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Acetoxy-2-phenylacetic Acid
CAS 5438-68-6
Molekylformel C10 H10 O4
Kvantitet 100 mg
Synonym Benzeneacetic acid, α-(acetyloxy)-, Mandelic acid, acetate (6CI,7CI,8CI), α-(Acetyloxy)benzeneacetic acid, (±)-O-Acetylmandelic acid, 2-(Acetyloxy)-2-phenylacetic acid, 2-Acetoxy-2-phenylacetic acid, Acetoxyphenylacetic acid, DL-O-Acetylmandelic acid, NSC
InChI formel InChI=1S/C10H10O4/c1-7(11)14-9(10(12)13)8-5-3-2-4-6-8/h2-6,9H,1H3,(H,12,13)
LEDER CC(=O)OC(C(=O)O)c1ccccc1
IUPAC-namn 2-acetyloxy-2-phenylacetic acid
Molekylvikt (g/mol) 194.184
Formel vikt 194.058