Läs mer
O-Acetyl Pseudoephedrine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (1S,2S)-O-Acetylpseudoephedrine Hydrochloride |
| CAS | 1630-34-8 |
| Molekylformel | C12 H17 N O2 . Cl H |
| Kvantitet | 100 mg |
| Synonym | Benzenemethanol, α-[1-(methylamino)ethyl]-, acetate (ester), hydrochloride, [S-(R*,R*)]- (9CI), Pseudoephedrine, acetate (ester), hydrochloride, (+)- (8CI), (1S,2S)-O-Acetylpseudoephedrine hydrochloride, (1S,2S)-2-(Methylamino)-1-phenylpropyl acetate hydrochloride, (αS)-α-[(1S)-1-(Methylamino)ethyl]benzenemethanol acetate (ester) hydrochloride |
| InChI formel | InChI=1S/C12H17NO2.ClH/c1-9(13-3)12(15-10(2)14)11-7-5-4-6-8-11;/h4-9,12-13H,1-3H3;1H/t9-,12+;/m0./s1 |
| LEDER | Cl.CN[C@@H](C)[C@@H](OC(=O)C)c1ccccc1 |
| IUPAC-namn | [(1S,2S)-2-(methylamino)-1-phenylpropyl] acetate;hydrochloride |
| Molekylvikt (g/mol) | 243.73 |
| Formel vikt | 243.1026 |